Learn More
Selleck Chemical LLC WAY-316606-2mg
SDP
Catalog No. 502389823
Encompass_Preferred
WAY-316606, a specific antagonist of SFRP1, functions as an activator of canonical Wnt/β-catenin signalling in the human hair bulb.
Catalog No. 50-238-9823 Supplier Selleck Chemical LLC Supplier No. S58152MG
WAY-316606, a specific antagonist of SFRP1, functions as an activator of canonical Wnt/β-catenin signalling in the human hair bulb.