Learn More
Medchemexpress LLC HY-10858 5mg Medchemexpress, WAY 316606 CAS:915759-45-4 Purity:>98%
SDP
Catalog No. 501968578
Encompass_Preferred
Medchemexpress, HY-10858 5mg WAY 316606 CAS:915759-45-4 WAY 316606 is an inhibitor of the secreted protein sFRP-1, an endogenous antagonist of the secreted glycoprotein Wnt. The affinity of WAY-316606 for sFRP-1 is determined using the FP binding assay with IC50 of 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8578 Supplier Medchemexpress LLC Supplier No. HY108585MG
Medchemexpress, HY-10858 5mg WAY 316606 CAS:915759-45-4 WAY 316606 is an inhibitor of the secreted protein sFRP-1, an endogenous antagonist of the secreted glycoprotein Wnt. The affinity of WAY-316606 for sFRP-1 is determined using the FP binding assay with IC50 of 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.