Learn More
Abcam WAY-316606, 1MG
Catalog No. 502680454
Encompass_Preferred
MW 448.5 Da, Purity >=98%. A secreted Frizzled-Related Protein-1 (sFRP-1) inhibitor (IC₅₀ = 0.65 μM). It blocks sFRP-1 from interacting with Wnt and thus increases Wnt signaling.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-0454 Supplier Abcam Supplier No. AB2871241MG
MW 448.5 Da, Purity >=98%. A secreted Frizzled-Related Protein-1 (sFRP-1) inhibitor (IC₅₀ = 0.65 μM). It blocks sFRP-1 from interacting with Wnt and thus increases Wnt signaling.
The product is subject to the following: Abcam Restricted Use Statement