Learn More
TARGETMOL CHEMICALS INC WAY-600 5MG
SDP
Catalog No. 502579299
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WAY-600 is a potent ATP-competitive and selective inhibitor of mTOR with IC50 of 9 nM; blocks mTORC1/P-S6K(T389) and mTORC2/P-AKT(S473) but not P-AKT(T308); selective for mTOR than PI3K(alpha) (>100-fold) and PI3K(gamma) (>500-fold). purity: 99%
Catalog No. 50-257-9299 Supplier TARGETMOL CHEMICALS INC Supplier No. T67305MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WAY-600 is a potent ATP-competitive and selective inhibitor of mTOR with IC50 of 9 nM; blocks mTORC1/P-S6K(T389) and mTORC2/P-AKT(S473) but not P-AKT(T308); selective for mTOR than PI3K(alpha) (>100-fold) and PI3K(gamma) (>500-fold). purity: 99%