Learn More
Apexbio Technology LLC WAY-600, 5mg. Cas: 1062159-35-6 MFCD: MFCD22419019. MTOR inhibitor
SDP
Supplier:  Apexbio Technology LLC B16415
Encompass_Preferred
WAY-600 is a potent, ATP-competitive and selective inhibitor of mTOR with IC50 of 9 nM; blocks mTORC1/P-S6K(T389) and mTORC2/P-AKT(S473) but not P-AKT(T308); selective for mTOR than PI3K (>100-fold) and PI3K (>500-fold). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5276