Learn More
Medchemexpress LLC (±)-WB 4101 hydrochloride | 2170-58-3 | 97.1% | 10 MM * 1 ML
SDP
Catalog No. 5000339245
Encompass_Preferred
(±)-WB 4101 hydrochloride is a potent antagonist of noradrenaline. It interacts with protein in smooth muscle, promoting tight binding between the drug and receptor.
  • Potent antagonist of noradrenaline
  • Interacts with protein in smooth muscle
  • Promotes tight binding between the drug and receptor
  • Store solid form at -20°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
Catalog No. 50-003-39245 Supplier Medchemexpress LLC Supplier No. HY12718A10MM/1ML
(±)-WB 4101 hydrochloride is a potent antagonist of noradrenaline. It interacts with protein in smooth muscle, promoting tight binding between the drug and receptor.
  • Potent antagonist of noradrenaline
  • Interacts with protein in smooth muscle
  • Promotes tight binding between the drug and receptor
  • Store solid form at -20°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture