Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WB-4101 hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013550 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3550 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3550 Supplier MilliporeSigma Supelco Supplier No. B018100MG
Add to Cart
Add to Cart

Specifications

Packaging Poly Bottle
CAS 2170-58-3
Physical Form Neat
Linear Formula C19H23 NO5 · HCl
MDL Number MFCD00055187
Synonym 2-[(2,6-Dimethoxyphenoxyethyl)aminomethyl]-1,4-benzodioxane hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 381.85
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.