Learn More
Cayman Chemical WB 4101hydrochlorIde 100mg
SDP
Catalog No. 502989080
Encompass_Preferred
An α-AR antagonist; inhibits norepinephrine-induced contraction in isolated rat vas deferens (pA2 = 9.8); inhibits carbamoylcholine-induced contractions in isolated guinea pig taenia caeci (pA2 = 8.9)
Catalog No. 50-298-9080 Supplier Cayman Chemical Supplier No. 29528100 MG
An α-AR antagonist; inhibits norepinephrine-induced contraction in isolated rat vas deferens (pA2 = 9.8); inhibits carbamoylcholine-induced contractions in isolated guinea pig taenia caeci (pA2 = 8.9)