Learn More
Medical Electronic Systems LLC QwikCheck WBC/pH Test Strips Quality Control, A-CA-01752-00, 12 months shelf life, 45 days opened shelf life, Colorimetric comparison, Room temperature storage
SDP
Catalog No. NC2172520
Encompass
QwikCheck WBC/pH Test Strips Quality Control, A-CA-01752-00, 12 months shelf life, 45 days opened shelf life, Colorimetric comparison, Room temperature storage
This item is not returnable. View return policy
Catalog No. NC2172520 Supplier Medical Electronic Systems LLC Supplier No. ACA0175200
QwikCheck WBC/pH Test Strips Quality Control, A-CA-01752-00, 12 months shelf life, 45 days opened shelf life, Colorimetric comparison, Room temperature storage