Learn More
MARSHALL SCIENTIFIC Polyscience WBE10A11B 10L Digital Water Bath 
SDP
Catalog No. NC2591249
Encompass_Preferred
The Polyscience WBE10A11B 10L Digital Water Bath is designed to handle a wide range of laboratory procedures including incubation, inactivation and agglutination as well as most pharmaceutical, serological, biomedical and industrial procedures. To ensure optimum temperature uniformity, your Water Bath features an energy efficient, large-area heater and thermostatic control. Its PID microprocessor control (Proportional Integral Derivative) system provides proportional heat control by anticipating the approach to your set point temperature and preventing overshoot. A redundant safety thermostat is standard on all models.
Catalog No. NC2591249 Supplier MARSHALL SCIENTIFIC Supplier No. WBE10A11B
The Polyscience WBE10A11B 10L Digital Water Bath is designed to handle a wide range of laboratory procedures including incubation, inactivation and agglutination as well as most pharmaceutical, serological, biomedical and industrial procedures. To ensure optimum temperature uniformity, your Water Bath features an energy efficient, large-area heater and thermostatic control. Its PID microprocessor control (Proportional Integral Derivative) system provides proportional heat control by anticipating the approach to your set point temperature and preventing overshoot. A redundant safety thermostat is standard on all models.