Learn More
Aviva Systems Biology Corp WBP11 Antibody - N-terminal region (ARP46241_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46241_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WBP11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WBP11. Alternate names: DKFZp779M1063, NPWBP, SIPP1, WBP-11ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59285