Learn More
Aviva Systems Biology Corp WBP2NL Antibody - middle region (ARP53072_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53072_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WBP2NL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WBP2NL. Alternate names: FLJ26145, MGC26816, PAWPApplicationWB. Species reactivity: Human
Catalog No. 50-014-60912