Learn More
Aviva Systems Biology Corp WBP4 Antibody - N-terminal region (ARP52293_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52293_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WBP4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human WBP4. Alternate names: FBP21, MGC117310ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-688