Learn More
Aviva Systems Biology Corp TCEAL9 Antibody - middle region (ARP39247_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39247_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WBP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human WBP5. Alternate names: DKFZp313K1940, TCEAL9ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-284-791