Learn More
Aviva Systems Biology Corp BUD23 Antibody - C-terminal region (ARP62733_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62733_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WBSCR22. It was validated on Western Blot. . Alternate names: HASJ4442, HUSSY-3, MGC19709, MGC2022, MGC5140, PP3381, WBMTApplicationWB. Species reactivity: Human
Catalog No. 50-287-758