Learn More
Medchemexpress LLC WCK-4234 | 1804915-68-1 | 98.0% | 269.21 g·mol⁻1 | C7H8N3NaO5S | 10MG
SDP
Catalog No. 5000236187
Encompass_Preferred
WCK-4234 is a research-grade β-lactamase inhibitor that targets class A, C, and D enzymes. It has no intrinsic antibacterial activity but enhances the effect of carbapenem antibiotics such as imipenem and meropenem against resistant Enterobacteriaceae, including strains with OXA-type carbapenemases. Intended for laboratory research use only.
  • Inhibits class A, C, and D β-lactamases.
  • Potentiates carbapenem activity against resistant Enterobacteriaceae.
  • Effective against OXA-type carbapenemase-mediated resistance.
  • Supplied with high purity suitable for biochemical and microbiological studies.
  • Molecular weight 269.21 g·mol⁻1 and formula C7H8N3NaO5S.
Catalog No. 50-002-36187 Supplier Medchemexpress LLC Supplier No. HY12560410MG
WCK-4234 is a research-grade β-lactamase inhibitor that targets class A, C, and D enzymes. It has no intrinsic antibacterial activity but enhances the effect of carbapenem antibiotics such as imipenem and meropenem against resistant Enterobacteriaceae, including strains with OXA-type carbapenemases. Intended for laboratory research use only.
  • Inhibits class A, C, and D β-lactamases.
  • Potentiates carbapenem activity against resistant Enterobacteriaceae.
  • Effective against OXA-type carbapenemase-mediated resistance.
  • Supplied with high purity suitable for biochemical and microbiological studies.
  • Molecular weight 269.21 g·mol⁻1 and formula C7H8N3NaO5S.