Learn More
Medchemexpress LLC WCK-4234 sodium | 1804915-68-1 | 98.0% | 269.21 g/mol | C7H8N3NaO5S | 5MG
SDP
Catalog No. 5000237304
Encompass_Preferred
WCK-4234 is a research-grade β-lactamase inhibitor used to study inhibition of class A, C, and D β-lactamases and to evaluate potentiation of carbapenem antibiotics. It has distinctive activity against OXA-type carbapenemases and is supplied as a sodium salt for laboratory use.
  • Potent β-lactamase inhibitor active against class A, C, and D enzymes.
  • Distinctive activity against OXA-type carbapenemases.
  • Potentiates carbapenem antibiotics in combination studies.
  • Supplied as a sodium salt with good solubility in DMSO.
  • High purity (>98%) suitable for research applications.
  • Available in small research-scale quantities for in vitro studies.
Catalog No. 50-002-37304 Supplier Medchemexpress LLC Supplier No. HY1256045MG
WCK-4234 is a research-grade β-lactamase inhibitor used to study inhibition of class A, C, and D β-lactamases and to evaluate potentiation of carbapenem antibiotics. It has distinctive activity against OXA-type carbapenemases and is supplied as a sodium salt for laboratory use.
  • Potent β-lactamase inhibitor active against class A, C, and D enzymes.
  • Distinctive activity against OXA-type carbapenemases.
  • Potentiates carbapenem antibiotics in combination studies.
  • Supplied as a sodium salt with good solubility in DMSO.
  • High purity (>98%) suitable for research applications.
  • Available in small research-scale quantities for in vitro studies.