Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore WC x LC Analytical Column, 10um, 4.6 x 250 mm, PEEK
Catalog No. NC2814218
Encompass_Preferred
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2814218 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B31110000004625P
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.