Learn More
NANOMICRO TECHNOLOGIES INC BioCore WCX LC Analytical Column, 5um, 4.6x150 mm, PEEK
Catalog No. NC2814229
Encompass_Preferred
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2814229 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B31105000004615P
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.