Learn More
NANOMICRO TECHNOLOGIES INC BioCore WCX LC Preparative Column, 5um, 10x250 mm, Stainless Steel
Catalog No. NC2814232
Encompass_Preferred
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC2814232 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B31105000010025S
Add to Cart
Add to Cart
Request a Quote
BioCore WCX is a family of high-performance, weak cation-exchange columns designed for separating charged variants in proteins, including monoclonal antibodies (mAbs). Its column technology involves the creation of a hydrophilic carboxylic functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.