Learn More
Aviva Systems Biology Corp WDFY1 Antibody - N-terminal region (ARP57465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDFY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WDFY1. Alternate names: FENS-1, WDF1, ZFYVE17ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62910