Learn More
Aviva Systems Biology Corp WDHD1 Antibody - middle region (ARP32844_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32844_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDHD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WDHD1. Alternate names: AND-1, CTF4, CHTF4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54792