Learn More
ABclonal Technology WDHD1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15396500UL
Encompass_Preferred
The protein encoded by this gene contains multiple N-terminal WD40 domains and a C-terminal high mobility group (HMG) box. WD40 domains are found in a variety of eukaryotic proteins and may function as adaptor/regulatory modules in signal transduction, pre-mRNA processing and cytoskeleton assembly. HMG boxes are found in many eukaryotic proteins involved in chromatin assembly, transcription and replication. Alternative splicing results in two transcript variants encoding different isoforms.
Catalog No. 50-001-04806