Learn More
Aviva Systems Biology Corp Wdr12 Antibody - middle region (ARP41316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Wdr12. It was validated on Western Blot. . Alternate names: 4933402C23Rik, Ytm1, Ytm1pApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-929