Learn More
Aviva Systems Biology Corp CFAP52 Antibody - middle region (ARP52794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR16. It was validated on Western Blot. WDR16,WD repeat domain 16,WDRPUH; FLJ37528; WDR16,WD repeat-containing protein 16,WD repeat-containing protein 16,WD40-repeat protein upregulated in HCC,WD40-repeat protein up-regulated in HCCThe immunogen is a synthetic peptide directed towards the middle region of human WDR16. Alternate names: FLJ37528, WDRPUHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-767