Learn More
Aviva Systems Biology Corp WDR34 Antibody - C-terminal region (ARP48430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR34. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human WDR34. Alternate names: MGC20486, RP11-216B9.5, bA216B9.3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59823