Learn More
Aviva Systems Biology Corp WDR35 Antibody - N-terminal region (ARP57449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WDR35. Alternate names: KIAA1336, MGC33196, CED2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53847