Learn More
Aviva Systems Biology Corp WDR4 Antibody - C-terminal region (ARP41321_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41321_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human WDR4. Alternate names: TRM82ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-57804