Learn More
Aviva Systems Biology Corp Wdr41 Antibody - middle region (ARP57186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Wdr41. It was validated on Western Blot. . Alternate names: B830029I03Rik, MSTP048ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-273