Learn More
Aviva Systems Biology Corp WDR49 Antibody - N-terminal region (ARP53376_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR49. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WDR49. Alternate names: FLJ33620ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62890