Learn More
Aviva Systems Biology Corp Wdr5 Antibody - C-terminal region (ARP41307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Wdr5. It was validated on Western Blot. . Alternate names: 2410008O07Rik, AA408785, AA960360, Big, Big-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-927