Learn More
Medchemexpress LLC WDR5 degrader-1 | 3032434-45-7 | 98.00% | 972.98 | 5 MG
SDP
Catalog No. 5000394928
Encompass_Preferred
WDR5 degrader-1 (compound 25) is a cereblon (CRBN)-recruiting WDR5 degrader. It selectively degrades WDR5 over the CRBN neo-substrate IKZF1.
  • Cereblon (CRBN)-recruiting degrader
  • Selectively degrades WDR5 over IKZF1
Catalog No. 50-003-94928 Supplier Medchemexpress LLC Supplier No. HY1620145MG
WDR5 degrader-1 (compound 25) is a cereblon (CRBN)-recruiting WDR5 degrader. It selectively degrades WDR5 over the CRBN neo-substrate IKZF1.
  • Cereblon (CRBN)-recruiting degrader
  • Selectively degrades WDR5 over IKZF1