Learn More
Sino Biological Recombinant Human KMT1A/SUV39H1 Protein (Full Length, GST Tag), Active
Catalog No. 502573055
Encompass_Preferred
Recombinant full length human SUV39H1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3055 Supplier Sino Biological Supplier No. S350380G50
Recombinant full length human SUV39H1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.