Learn More
Aviva Systems Biology Corp WDR51B Antibody - N-terminal region (ARP55620_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55620_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR51B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WDR51B. Alternate names: FLJ14923, FLJ41111, TUWD12, PIX1, WDR51BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61555