Learn More
Aviva Systems Biology Corp WDR53 Antibody - middle region (ARP55842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR53. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WDR53. Alternate names: MGC64882ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61630