Learn More
Aviva Systems Biology Corp WDR66 Antibody - N-terminal region (ARP52729_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52729_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR66. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WDR66. Alternate names: FLJ39783, MGC33630ApplicationWB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-60814