Learn More
Aviva Systems Biology Corp WDR73 Antibody - middle region (ARP60698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR73. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human WDR73. Alternate names: FLJ14888, HSPC264ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-287-131