Learn More
Aviva Systems Biology Corp Wdr8 Antibody - N-terminal region (ARP41308_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41308_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Wdr8. It was validated on Western Blot. . Alternate names: 2610044M17Rik, 5330425N03Rik, Dd57, Wrap73, Wdr8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-928