Learn More
Aviva Systems Biology Corp WDR8 Antibody - middle region (ARP41309_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41309_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDR8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WDR8. Alternate names: FLJ20430, MGC99569, WDR8ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-57799