Learn More
MICRO EPSILON AMERICA WDS1500P60SRI D-W D SENSOR
Catalog No. NC2147342
Encompass
WDS-1500-P60-SR-I Draw-wire displacement sensormeasuring range 1500 mm,model P60,linearity +/- 0,1% FSO,power supply 14 - 27 Vdc,non stabilized,output current 4... 20 mA,protection class IP 65,flange connector, radial, 8-pinincl. mating connector
Catalog No. NC2147342 Supplier MICRO EPSILON AMERICA Supplier No. 2625100
Add to Cart
Add to Cart
WDS-1500-P60-SR-I Draw-wire displacement sensormeasuring range 1500 mm,model P60,linearity +/- 0,1% FSO,power supply 14 - 27 Vdc,non stabilized,output current 4... 20 mA,protection class IP 65,flange connector, radial, 8-pinincl. mating connector