Learn More
Aviva Systems Biology Corp WDSUB1 Antibody - middle region (ARP43423_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43423_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WDSUB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WDSUB1. Alternate names: FLJ36175, UBOX6, WDSAM1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58528