Learn More
MSC Wearwell Anti-Fatigue Mat, 10 ft. Long, 1 ea
Catalog No. 5000675133
Encompass
This anti-fatigue mat is designed for heavy-duty use in dry environments, providing comfort and reducing fatigue for workers. It features a diamond plate surface for enhanced traction and beveled edges to prevent tripping hazards. Constructed from a composite of vinyl sponge and nitrile rubber, it offers durability and resistance to chemicals and cutting fluids.
  • Heavy-duty traffic type
  • Vinyl surface and vinyl sponge/nitrile rubber base material
  • Thickness of 9/16 in.
  • Length of 10 ft., width of 3 ft.
  • Diamond plate surface pattern
  • Beveled edges for safety
  • Chemical and cutting fluid-resistant
  • Suitable for dry environments
  • Meets ASTM D575, ASTM F1677, MVSS 302, and Federal Standard 191
  • Made from recycled content
Catalog No. 50-006-75133 Supplier MSC Supplier No. 79460416
This anti-fatigue mat is designed for heavy-duty use in dry environments, providing comfort and reducing fatigue for workers. It features a diamond plate surface for enhanced traction and beveled edges to prevent tripping hazards. Constructed from a composite of vinyl sponge and nitrile rubber, it offers durability and resistance to chemicals and cutting fluids.
  • Heavy-duty traffic type
  • Vinyl surface and vinyl sponge/nitrile rubber base material
  • Thickness of 9/16 in.
  • Length of 10 ft., width of 3 ft.
  • Diamond plate surface pattern
  • Beveled edges for safety
  • Chemical and cutting fluid-resistant
  • Suitable for dry environments
  • Meets ASTM D575, ASTM F1677, MVSS 302, and Federal Standard 191
  • Made from recycled content