Learn More
MSC Wearwell Anti-Fatigue Mat, 18x18 in, Black, 1 ea
Catalog No. 5000684320
Encompass
This solid grid mat is designed for dry environments and heavy-duty traffic, providing standing relief. Constructed from a durable vinyl base, it offers resistance to abrasion, chemicals, and cutting fluids.
  • Designed for dry applications
  • Customizable in 3-inch increments to fit any area
  • Positive interlocking system prevents separation
  • Easy replacement of single pieces or sections to reduce costs
  • Made from 100% silicone-free PVC tiles for maximum durability
  • Made from recycled content
  • Abrasion-resistant
  • Chemical-resistant
  • Cutting fluid-resistant
  • Features 4 interlocking sides
Catalog No. 50-006-84320 Supplier MSC Supplier No. 75330613
This solid grid mat is designed for dry environments and heavy-duty traffic, providing standing relief. Constructed from a durable vinyl base, it offers resistance to abrasion, chemicals, and cutting fluids.
  • Designed for dry applications
  • Customizable in 3-inch increments to fit any area
  • Positive interlocking system prevents separation
  • Easy replacement of single pieces or sections to reduce costs
  • Made from 100% silicone-free PVC tiles for maximum durability
  • Made from recycled content
  • Abrasion-resistant
  • Chemical-resistant
  • Cutting fluid-resistant
  • Features 4 interlocking sides