Learn More
MSC Wearwell Anti-Fatigue Mat, 20 ft x 3 ft, 1 ea
Catalog No. 5000688142
Encompass
This anti-fatigue mat is designed for demanding industrial environments. It measures 20 ft long, 3 ft wide, and 1/2 inch thick, and is constructed from durable CFR rubber. Featuring beveled edges, this mat is suitable for heavy-duty traffic in both dry and wet areas.
  • Measures 20 ft in length and 3 ft in width
  • Features a 1/2 in thickness
  • Constructed from CFR rubber for durability
  • Designed with beveled edges for safety
  • Suitable for heavy-duty traffic applications
  • Effective in both dry and wet environments
  • Features a raised circle surface pattern
  • Resistant to chemicals and cutting fluids
  • Includes drainage holes
  • Made from recycled content
Catalog No. 50-006-88142 Supplier MSC Supplier No. 85531713
This anti-fatigue mat is designed for demanding industrial environments. It measures 20 ft long, 3 ft wide, and 1/2 inch thick, and is constructed from durable CFR rubber. Featuring beveled edges, this mat is suitable for heavy-duty traffic in both dry and wet areas.
  • Measures 20 ft in length and 3 ft in width
  • Features a 1/2 in thickness
  • Constructed from CFR rubber for durability
  • Designed with beveled edges for safety
  • Suitable for heavy-duty traffic applications
  • Effective in both dry and wet environments
  • Features a raised circle surface pattern
  • Resistant to chemicals and cutting fluids
  • Includes drainage holes
  • Made from recycled content