Learn More
MSC Wearwell Anti-Fatigue Mat, 5x3 ft, 3/8 in, 1 ea
Catalog No. 5000766338
Encompass
This anti-fatigue mat is designed for light-duty traffic in dry environments, providing practical and economical comfort. It is constructed from a blown PVC sponge, which can reduce floor hardness by up to 50%, and features safety rounded edges. The mat can be custom cut to fit specific work areas.
  • Economic comfort for light industrial applications
  • Safety rounded edges
  • Easily cut to size on-site
  • Overall thickness 3/8 in
  • Made from recycled content
  • Suitable for dry floors with occasional spills
  • Ribbed surface pattern
  • Base material is vinyl sponge
  • Surface material is vinyl
Catalog No. 50-007-66338 Supplier MSC Supplier No. 89863138
This anti-fatigue mat is designed for light-duty traffic in dry environments, providing practical and economical comfort. It is constructed from a blown PVC sponge, which can reduce floor hardness by up to 50%, and features safety rounded edges. The mat can be custom cut to fit specific work areas.
  • Economic comfort for light industrial applications
  • Safety rounded edges
  • Easily cut to size on-site
  • Overall thickness 3/8 in
  • Made from recycled content
  • Suitable for dry floors with occasional spills
  • Ribbed surface pattern
  • Base material is vinyl sponge
  • Surface material is vinyl