Learn More
MSC Wearwell Anti-Fatigue Mat, 5x3 ft, Vinyl, 1 ea
Catalog No. 5000793672
Encompass
This anti-fatigue mat is 5 ft long and 3 ft wide with a 9/16 in thickness. Constructed from a vinyl sponge base and vinyl surface, it features a diamond plate pattern with beveled edges, ideal for dry, medium-duty industrial environments.
  • Composite construction
  • Abrasion-resistant
  • Resistant to chemicals and cutting fluids
  • Meets ASTM F1677, ASTM D395-98, MVSS 302, and Federal Standard 191 standards
  • Made from recycled content
Catalog No. 50-007-93672 Supplier MSC Supplier No. 48445308
This anti-fatigue mat is 5 ft long and 3 ft wide with a 9/16 in thickness. Constructed from a vinyl sponge base and vinyl surface, it features a diamond plate pattern with beveled edges, ideal for dry, medium-duty industrial environments.
  • Composite construction
  • Abrasion-resistant
  • Resistant to chemicals and cutting fluids
  • Meets ASTM F1677, ASTM D395-98, MVSS 302, and Federal Standard 191 standards
  • Made from recycled content