Learn More
MSC Wearwell Anti-Fatigue Mat, 60 x 24 in, 1 ea
Catalog No. 5000793630
Encompass
Wearwell Anti-Fatigue Modular Tiles Mats are designed for dry environments to reduce fatigue for workers. This black mat measures 60 in long, 24 in wide, and 5/8 in thick.
  • Urethane and vinyl sponge base
  • Raised squares and domes surface style
  • Drainage holes
  • Abrasion-resistant
  • Cutting fluid-resistant
  • Complies with ASTM D 395 and ASTM F1677 standards
  • Made from recycled content
Catalog No. 50-007-93630 Supplier MSC Supplier No. 74166372
Wearwell Anti-Fatigue Modular Tiles Mats are designed for dry environments to reduce fatigue for workers. This black mat measures 60 in long, 24 in wide, and 5/8 in thick.
  • Urethane and vinyl sponge base
  • Raised squares and domes surface style
  • Drainage holes
  • Abrasion-resistant
  • Cutting fluid-resistant
  • Complies with ASTM D 395 and ASTM F1677 standards
  • Made from recycled content