Learn More
MSC Wearwell Anti-Fatigue Mat, Heavy-Duty, 60 x 36 in, 1 ea
Catalog No. 5000793481
Encompass
This anti-fatigue mat is engineered for heavy-duty applications in dry environments. Featuring a composite construction with a vinyl surface and nitrile rubber base, it provides comfort and support, reducing fatigue for workers.
  • Beveled edges for safety
  • Diamond plate surface with grit coating for enhanced traction
  • Chemical and cutting fluid-resistant
  • Made from recycled content
  • Complies with ASTM D575, ASTM F1677, MVSS 302, and Federal Standard 191
Catalog No. 50-007-93481 Supplier MSC Supplier No. 75330712
This anti-fatigue mat is engineered for heavy-duty applications in dry environments. Featuring a composite construction with a vinyl surface and nitrile rubber base, it provides comfort and support, reducing fatigue for workers.
  • Beveled edges for safety
  • Diamond plate surface with grit coating for enhanced traction
  • Chemical and cutting fluid-resistant
  • Made from recycled content
  • Complies with ASTM D575, ASTM F1677, MVSS 302, and Federal Standard 191