Learn More
MSC Wearwell Anti-Fatigue Mat, Vinyl, 1 ea
Catalog No. 5000677094
Encompass
This anti-fatigue mat provides comfort and support in dry work environments. Designed for medium-duty traffic, it features a durable vinyl construction with a diamond plate surface.
  • Measuring 8 ft. × 3 ft.
  • With 5/8 in. thickness
  • Beveled edges
  • Vinyl sponge base with vinyl surface
  • Composite construction
  • Abrasion-resistant
  • Suitable for dry environments
  • Two colored borders
  • Made from recycled content
Catalog No. 50-006-77094 Supplier MSC Supplier No. 32615825
This anti-fatigue mat provides comfort and support in dry work environments. Designed for medium-duty traffic, it features a durable vinyl construction with a diamond plate surface.
  • Measuring 8 ft. × 3 ft.
  • With 5/8 in. thickness
  • Beveled edges
  • Vinyl sponge base with vinyl surface
  • Composite construction
  • Abrasion-resistant
  • Suitable for dry environments
  • Two colored borders
  • Made from recycled content