Learn More
MSC Wearwell Edging, Female, Grease Resistant, Yellow, 1 ea
Catalog No. 5000787731
Encompass
This 24/Seven GR (Grease Resistant) female edging securely interlocks with 24/Seven GR rubber floor tiles. The yellow edging serves as a visual cue and provides a smooth transition for foot traffic in areas with water or dry debris.
  • Grease resistant (GR) rubber construction
  • Provides a visual cue and smooth transition for foot traffic
  • Ideal for areas with water or dry debris, wet or dry floors, weak acids, and alcohols
  • Measures 3 in (8 cm) wide, 39 in (99 cm) long, and 5/8 in (16 mm) thick
  • Suitable for manufacturing assembly lines, airports, workshops, garages, and warehouses
Catalog No. 50-007-87731 Supplier MSC Supplier No. 96555602
This 24/Seven GR (Grease Resistant) female edging securely interlocks with 24/Seven GR rubber floor tiles. The yellow edging serves as a visual cue and provides a smooth transition for foot traffic in areas with water or dry debris.
  • Grease resistant (GR) rubber construction
  • Provides a visual cue and smooth transition for foot traffic
  • Ideal for areas with water or dry debris, wet or dry floors, weak acids, and alcohols
  • Measures 3 in (8 cm) wide, 39 in (99 cm) long, and 5/8 in (16 mm) thick
  • Suitable for manufacturing assembly lines, airports, workshops, garages, and warehouses